Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
dctd
Entrez Gene ID
549019
Gene Name
dCMP deaminase
Gene Aliases
-
Chromosome Location
Chr.1: 36232726 - 36240438 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016265.2 NP_001016265.1 5 - 6 502 103
GenBank mRNA CR761692.2 - 5 - 6 502 103

View Full Product Details