Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
casp7
Entrez Gene ID
549053
Gene Name
caspase 7
Gene Aliases
caspase-7, caspase7, cmh-1, ice-lap3, mch3
Chromosome Location
Chr.Un|NW_016683886: 50204 - 55063 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016299.2 NP_001016299.1 4 - 5 759 102
GenBank mRNA BC170775.1 - 4 - 5 757 102
BC170801.1 - 4 - 5 757 102
CR761526.2 - 4 - 5 759 102

View Full Product Details