Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
edf1
Entrez Gene ID
549440
Gene Name
endothelial differentiation related factor 1
Gene Aliases
edf-1, mbf1
Chromosome Location
Chr.8: 369258 - 374488 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016686.2 NP_001016686.1 1 - 2 95 88
GenBank mRNA CR760196.2 - 1 - 2 95 88

View Full Product Details