Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rsph6a
Entrez Gene ID
549514
Gene Name
radial spoke head 6 homolog A
Gene Aliases
rshl1, rsp4, rsp6, rsph4b
Chromosome Location
Chr.8: 49887351 - 49898443 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001016760.2 NP_001016760.1 2 - 3 392 74
XM_012968440.1 XP_012823894.1 4 - 5 481 74
GenBank mRNA BC127351.1 - 2 - 3 359 74
CR855771.2 - 2 - 3 392 74

View Full Product Details