Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mrpl32
Entrez Gene ID
549991
Gene Name
mitochondrial ribosomal protein L32
Gene Aliases
-
Chromosome Location
Chr.6: 66004945 - 66008792 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001017237.2 NP_001017237.1 2 - 3 322 91
GenBank mRNA CR760203.2 - 2 - 3 322 91

View Full Product Details