Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
yipf6
Entrez Gene ID
548686
Gene Name
Yip1 domain family member 6
Gene Aliases
-
Chromosome Location
Chr.8: 20100883 - 20109185 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001015932.3 NP_001015932.1 5 - 6 430 98
XM_018096248.1 XP_017951737.1 4 - 5 399 98
GenBank mRNA BC122940.1 - 5 - 6 430 98
CR926375.2 - 5 - 6 426 98

View Full Product Details