Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
psmc1
Entrez Gene ID
733980
Gene Name
proteasome 26S subunit, ATPase 1
Gene Aliases
p26s4, p56, pros26.4
Chromosome Location
Chr.8: 65646815 - 65656451 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045689.2 NP_001039154.1 2 - 3 132 106
GenBank mRNA CR942586.2 - 1 - 2 61 106

View Full Product Details