Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
lancl2
Entrez Gene ID
734145
Gene Name
LanC like 2
Gene Aliases
-
Chromosome Location
Chr.6: 65566178 - 65594195 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001045803.1 NP_001039268.1 2 - 3 304 91
XM_012964858.2 XP_012820312.1 3 - 4 515 91
XM_012964859.2 XP_012820313.1 4 - 5 653 91
XM_018094689.1 XP_017950178.1 4 - 5 581 91
GenBank mRNA BC135988.1 - 4 - 5 631 91
CR942790.2 - 2 - 3 304 91

View Full Product Details