Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
awat1
Entrez Gene ID
100127622
Gene Name
acyl-CoA wax alcohol acyltransferase 1
Gene Aliases
dga2, dgat2l3
Chromosome Location
Chr.5: 121571591 - 121584105 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001112975.1 NP_001106446.1 6 - 7 683 81
XM_012969998.2 XP_012825452.1 6 - 7 1366 81
GenBank mRNA BC153737.1 - 6 - 7 683 81

View Full Product Details