Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ptar1
Entrez Gene ID
100145268
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
ptar1-a, ptar1-b
Chromosome Location
Chr.1: 117364913 - 117394097 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001126747.1 NP_001120219.1 5 - 6 647 62
GenBank mRNA BC159397.1 - 5 - 6 647 62

View Full Product Details