Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fundc1
Entrez Gene ID
100145592
Gene Name
FUN14 domain containing 1
Gene Aliases
-
Chromosome Location
Chr.2: 69113744 - 69120834 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127004.1 NP_001120476.1 4 - 5 378 95
GenBank mRNA BC161330.1 - 4 - 5 378 95

View Full Product Details