Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
rarres1
Entrez Gene ID
100158540
Gene Name
retinoic acid receptor responder (tazarotene induced) 1
Gene Aliases
tig1
Chromosome Location
Chr.5: 103800554 - 103825215 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001127974.1 NP_001121446.1 4 - 5 684 96
GenBank mRNA BC166204.1 - 4 - 5 684 96

View Full Product Details