Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
abhd11
Entrez Gene ID
779475
Gene Name
abhydrolase domain containing 11
Gene Aliases
pp1226, wbscr21
Chromosome Location
Chr.2: 38151771 - 38160210 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001128647.2 NP_001122119.2 4 - 5 497 79
XM_018091329.1 XP_017946818.1 5 - 6 591 79
GenBank mRNA BC121505.1 - 4 - 5 497 79
BC167663.1 - 4 - 5 498 79

View Full Product Details