Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
btbd1
Entrez Gene ID
100170424
Gene Name
BTB (POZ) domain containing 1
Gene Aliases
ns5atp8
Chromosome Location
Chr.3: 95461804 - 95506435 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130202.1 NP_001123674.1 4 - 5 635 76
XM_018091949.1 XP_017947438.1 5 - 6 955 76
GenBank mRNA BC166965.1 - 4 - 5 635 76

View Full Product Details