Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rgs16
Entrez Gene ID
100170622
Gene Name
regulator of G-protein signaling 16
Gene Aliases
-
Chromosome Location
Chr.4: 90037493 - 90041066 on Build UCB_Xtro_10.0
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130383.1 NP_001123855.1 1 - 2 54 79
GenBank mRNA BC167734.1 - 1 - 2 54 79

View Full Product Details