Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
LOC100281809
Entrez Gene ID
100281809
Gene Name
protein kinase APK1B
Gene Aliases
-
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001154729.1 NP_001148201.1 - 1441 65
GenBank mRNA BT062751.1 - - 1437 65
EU958039.1 - - 1441 65

Gene Symbol
LOC100381452
Entrez Gene ID
100381452
Gene Name
putative protein kinase superfamily protein
Gene Aliases
ZEAMMB73_319340
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001174290.1 NP_001167761.1 - 1357 65
GenBank mRNA BT061299.1 - - 1357 65

View Full Product Details