Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
cys2
Entrez Gene ID
542438
Gene Name
cysteine synthase2
Gene Aliases
Mcysp
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111999.1 NP_001105469.1 - 225 56
GenBank mRNA X85803.1 - - 225 56

View Full Product Details