Assay Design

The assay is designed to a sequence that does not exhibit typical exon-intron-exon gene structure, such as a fusion transcript, or an artificial, prokaryotic or mitochondrial sequence.

Gene Transcripts

Gene Symbol
m4
Entrez Gene ID
542041
Gene Name
m4 protein
Gene Aliases
ZEAMMB73_053100
Chromosome Location
-
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001111681.1 NP_001105151.1 - 936 102
GenBank mRNA AJ430641.1 - - 936 102
BT085681.1 - - 940 102

View Full Product Details