Gene Transcripts
- Gene Symbol
- Vax2os
- Entrez Gene ID
- 574519
- Gene Name
- ventral anterior homeobox 2, opposite strand
- Gene Aliases
- Vas2os, Vax2os1, Vax2os2
- Chromosome Location
- Chr. 6: 83692806 - 83712201 on Build GRCm38
- UniGene ID
- Mm.487605
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_002871.1 |
|
6 |
1178 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BB667171.1 |
- |
|
396 |
|
Chr. - - [-] |
View Full Product Details