Gene Transcripts
- Gene Symbol
- MYADML
- Entrez Gene ID
- 151325
- Gene Name
- myeloid associated differentiation marker like (pseudogene)
- Gene Aliases
- Chromosome Location
- Chr. 2: 33726061 - 33728217 on Build GRCh38
- UniGene ID
- Hs.528847
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003143.2 |
|
1 |
1927 |
|
GenBank mRNA |
AK057470.1 |
- |
1 |
1920 |
|
BC031088.1 |
- |
1 |
1923 |
|
View Full Product Details