Gene Transcripts
- Gene Symbol
- SNORA79
- Entrez Gene ID
- 677845
- Gene Name
- small nucleolar RNA, H/ACA box 79
- Gene Aliases
- ACA65, SNORA79A
- Chromosome Location
- Chr. 14: 81202695 - 81202834 on Build GRCh38
- UniGene ID
- Hs.693251
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_003021.2 |
|
1 |
102 |
|
View Full Product Details