Gene Transcripts
- Gene Symbol
- LINC00520
- Entrez Gene ID
- 645687
- Gene Name
- long intergenic non-protein coding RNA 520
- Gene Aliases
- C14orf34
- Chromosome Location
- Chr. 14: 55781135 - 55796674 on Build GRCh38
- UniGene ID
- Hs.569345
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026796.1 |
|
4 |
832 |
|
NR_026797.1 |
|
3 |
770 |
|
View Full Product Details