Gene Transcripts
- Gene Symbol
- ZNF252P-AS1
- Entrez Gene ID
- 286103
- Gene Name
- ZNF252P antisense RNA 1
- Gene Aliases
- C8orf77
- Chromosome Location
- Chr. 8: 145002811 - 145006046 on Build GRCh38
- UniGene ID
- Hs.359978
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_026974.1 |
|
1 |
2733 |
|
GenBank mRNA |
AK092777.1 |
- |
1 |
2558 |
|
BC046439.1 |
- |
1 |
2733 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ185413.1 |
- |
1 |
340 |
|
Chr. 8 - 145005204 - 145005938 [+] |
View Full Product Details