Gene Transcripts
- Gene Symbol
- KLHL9
- Entrez Gene ID
- 55958
- Gene Name
- kelch like family member 9
- Gene Aliases
- Chromosome Location
- Chr. 9: 21331019 - 21335430 on Build GRCh38
- UniGene ID
- Hs.701778
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK022805.1 |
- |
1 |
4401 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
N99347.1 |
- |
2 |
1979 |
|
Chr. 9 - 21324052 - 21326339 [-] |
View Full Product Details