Gene Transcripts
- Gene Symbol
- CRHR1
- Entrez Gene ID
- 1394
- Gene Name
- corticotropin releasing hormone receptor 1
- Gene Aliases
- CRF-R, CRF-R-1, CRF-R1, CRF1, CRFR-1, CRFR1, CRH-R-1, CRH-R1, CRHR, CRHR1L
- Chromosome Location
- Chr. 17: 45784280 - 45835828 on Build GRCh38
- UniGene ID
- Hs.417628
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK124894.1 |
- |
2 |
2800 |
|
View Full Product Details