Gene Transcripts
- Gene Symbol
- EBF2
- Entrez Gene ID
- 64641
- Gene Name
- early B-cell factor 2
- Gene Aliases
- COE2, EBF-2, O/E-3, OE-3
- Chromosome Location
- Chr. 8: 25841730 - 26045124 on Build GRCh38
- UniGene ID
- Hs.491292
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW962848.1 |
- |
1 |
200 |
|
Chr. 8 - 25842051 - 25842540 [-] |
GenBankmRNA |
AA311763.1 |
- |
1 |
201 |
|
Chr. 8 - 25842075 - 25842541 [-] |
View Full Product Details