Gene Transcripts

Gene Symbol
ANKRD44
Entrez Gene ID
91526
Gene Name
ankyrin repeat domain 44
Gene Aliases
PP6-ARS-B
Chromosome Location
Chr. 2: 196962617 - 197310797 on Build GRCh38
UniGene ID
Hs.432706
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001195144.1 NP_001182073.1 28 5238
XM_005246945.1 XP_005247002.1 28 6560
XM_005246946.1 XP_005247003.1 28 6506
XM_006712838.3 XP_006712901.1 28 6729
XM_011512148.2 XP_011510450.1 28 8441
XM_017005252.1 XP_016860741.1 28 8387
GenBank mRNA AK021632.1 - 1 1076

View Full Product Details