Gene Transcripts

Gene Symbol
ZNF99
Entrez Gene ID
7652
Gene Name
zinc finger protein 99
Gene Aliases
C19orf9, F8281
Chromosome Location
Chr. 19: 22752183 - 22784171 on Build GRCh38
UniGene ID
Hs.568380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001080409.2 NP_001073878.2 4 6172
GenBank mRNA BC021822.1 - 1 984

View Full Product Details