Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001290046.1 | NP_001276975.1 | 19 | 9594 | |
NM_001290047.1 | NP_001276976.1 | 19 | 10066 | ||
XM_006724079.3 | XP_006724142.1 | 7 | 8509 | ||
XM_011546128.2 | XP_011544430.1 | 18 | 9670 | ||
XM_011546129.2 | XP_011544431.1 | 19 | 9670 | ||
XM_011546132.2 | XP_011544434.1 | 19 | 9589 | ||
XM_017028785.1 | XP_016884274.1 | 19 | 9751 | ||
GenBank mRNA | AK055696.1 | - | 1 | 1541 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GenBankmRNA | BQ950712.1 | - | 1 | 725 | Chr. 22 - 17557370 - 17558142 [+] |