Gene Transcripts
- Gene Symbol
- GPI
- Entrez Gene ID
- 2821
- Gene Name
- glucose-6-phosphate isomerase
- Gene Aliases
- AMF, GNPI, NLK, PGI, PHI, SA-36, SA36
- Chromosome Location
- Chr. 19: 34359592 - 34402413 on Build GRCh38
- UniGene ID
- Hs.466471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX641158.1 |
- |
1 |
958 |
|
View Full Product Details