Gene Transcripts
- Gene Symbol
- UBE2G2
- Entrez Gene ID
- 7327
- Gene Name
- ubiquitin conjugating enzyme E2 G2
- Gene Aliases
- UBC7
- Chromosome Location
- Chr. 21: 44768580 - 44801836 on Build GRCh38
- UniGene ID
- Hs.529420
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC054341.1 |
- |
1 |
278 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BQ424505.1 |
- |
1 |
286 |
|
Chr. 21 - 44801004 - 44801794 [-] |
View Full Product Details