Gene Transcripts

Gene Symbol
UCHL3
Entrez Gene ID
7347
Gene Name
ubiquitin C-terminal hydrolase L3
Gene Aliases
UCH-L3
Chromosome Location
Chr. 13: 75549480 - 75606020 on Build GRCh38
UniGene ID
Hs.162241
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020726.1 XP_016876215.1 9 1974
GenBank mRNA AK093882.1 - 1 2066

View Full Product Details