Gene Transcripts
- Gene Symbol
- Golm1
- Entrez Gene ID
- 105348
- Gene Name
- golgi membrane protein 1
- Gene Aliases
- 2310001L02Rik, AW125446, D030064E01Rik, GP73, Golph2, PSEC0257
- Chromosome Location
- Chr. 13: 59634996 - 59675784 on Build GRCm38
- UniGene ID
- Mm.171335
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK051066.1 |
- |
1 |
1720 |
|
View Full Product Details