Gene Transcripts
- Gene Symbol
- Trpm3
- Entrez Gene ID
- 226025
- Gene Name
- transient receptor potential cation channel, subfamily M, member 3
- Gene Aliases
- 6330504P12Rik, 9330180E14, AU018608, B930001P07Rik, LTRPC3, MLSN2
- Chromosome Location
- Chr. 19: 22138181 - 22989897 on Build GRCm38
- UniGene ID
- Mm.131943
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK031944.1 |
- |
|
106 |
|
AK034340.1 |
- |
|
334 |
|
AK139989.1 |
- |
|
1817 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CN458801.1 |
- |
1 |
147 |
|
Chr. 19 - 22992260 - 22992903 [+] |
View Full Product Details