Gene Transcripts
- Gene Symbol
- Impad1
- Entrez Gene ID
- 242291
- Gene Name
- inositol monophosphatase domain containing 1
- Gene Aliases
- 1110001C20Rik, AA408880, AI451589, AL022796, B230207P20, Jaws, gPAPP
- Chromosome Location
- Chr. 4: 4764342 - 4793306 on Build GRCm38
- UniGene ID
- Mm.218889
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK003208.1 |
- |
1 |
660 |
|
AK041202.1 |
- |
|
3361 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BY703521.1 |
- |
1 |
654 |
|
Chr. 4 - 4764032 - 4764696 [-] |
GenBankmRNA |
BB817280.1 |
- |
1 |
384 |
|
Chr. 4 - 4764032 - 4764426 [-] |
GenBankmRNA |
CK636528.1 |
- |
1 |
471 |
|
Chr. 4 - 4764031 - 4764512 [-] |
View Full Product Details