Gene Transcripts
- Gene Symbol
- RGS5
- Entrez Gene ID
- 8490
- Gene Name
- regulator of G-protein signaling 5
- Gene Aliases
- MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129
- Chromosome Location
- Chr. 1: 163142299 - 163321791 on Build GRCh38
- UniGene ID
- Hs.24950
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_045630.1 |
|
4 |
504 |
|
GenBank mRNA |
BC032239.1 |
- |
4 |
465 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000439699.1 |
- |
4 |
465 |
|
Chr. 1 - 163304211 - 163321752 [-] |
View Full Product Details