Gene Transcripts
- Gene Symbol
- SNHG3
- Entrez Gene ID
- 8420
- Gene Name
- small nucleolar RNA host gene 3
- Gene Aliases
- NCRNA00014, RNU17C, RNU17D, U17HG, U17HG-A, U17HG-AB
- Chromosome Location
- Chr. 1: 28505943 - 28510892 on Build GRCh38
- UniGene ID
- Hs.469723
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK092726.1 |
- |
1 |
1008 |
|
- Gene Symbol
- SNORA73A
- Entrez Gene ID
- 6080
- Gene Name
- small nucleolar RNA, H/ACA box 73A
- Gene Aliases
- E1, E1-7, E1b, RNE1, RNU17A, U17A
- Chromosome Location
- Chr. 1: 28507365 - 28507571 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
CX783521.1 |
- |
1 |
491 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GENCODE |
ENST00000437681.1 |
- |
1 |
1008 |
|
Chr. 1 - 28506351 - 28509633 [+] |
View Full Product Details