Gene Transcripts

Gene Symbol
Krtap19-1
Entrez Gene ID
170657
Gene Name
keratin associated protein 19-1
Gene Aliases
Krtap16-2, Krtap16-9
Chromosome Location
Chr. 16: 88868918 - 88869410 on Build GRCm38
UniGene ID
Mm.196822
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_130876.3 NP_570946.2 1 357
GenBank mRNA AA930779.1 - 1 49
AF345292.1 - 1 314

View Full Product Details