Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001289729.1 | NP_001276658.1 | 18 | 2276 | |
NM_133213.2 | NP_573476.2 | 18 | 2075 | ||
GenBank mRNA | AF367247.1 | - | 18 | 1739 | |
AF428102.1 | - | 18 | 1926 | ||
AK078835.1 | - | 18 | 1882 | ||
AK085520.1 | - | 18 | 2074 | ||
AK087627.1 | - | 18 | 1882 | ||
AK164538.1 | - | 18 | 1903 | ||
AK172086.1 | - | 18 | 2076 | ||
BC140977.1 | - | 18 | 1719 | 9055589 |