Gene Transcripts

Gene Symbol
Evi5l
Entrez Gene ID
213027
Gene Name
ecotropic viral integration site 5 like
Gene Aliases
1700084G18Rik, 2310039H16Rik, 3110007G05Rik, B130050I23Rik
Chromosome Location
Chr. 8: 4166538 - 4208050 on Build GRCm38
UniGene ID
Mm.437106
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001039578.3 NP_001034667.3 3 554
GenBank mRNA AK045240.1 - 3 555
AK087482.1 - 3 555
BC042653.1 - 3 534

View Full Product Details