Gene Transcripts

Gene Symbol
Lingo3
Entrez Gene ID
237403
Gene Name
leucine rich repeat and Ig domain containing 3
Gene Aliases
AI851712, LERN2
Chromosome Location
Chr. 10: 80832801 - 80844039 on Build GRCm38
UniGene ID
Mm.40213
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013758.2 NP_001013780.2 2 1818
GenBank mRNA BC072620.1 - 2 1809

View Full Product Details