Gene Transcripts

Gene Symbol
Morn2
Entrez Gene ID
378462
Gene Name
MORN repeat containing 2
Gene Aliases
Mopt
Chromosome Location
Chr. 17: 80290212 - 80305691 on Build GRCm38
UniGene ID
Mm.45208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_194269.2 NP_919245.1 5 448
GenBank mRNA AY367765.1 - 5 448
BC037365.1 - 4 374
BC089486.1 - 3 223 6742814

View Full Product Details