Gene Transcripts

Gene Symbol
Tlcd2
Entrez Gene ID
380712
Gene Name
TLC domain containing 2
Gene Aliases
Chromosome Location
Chr. 11: 75468024 - 75470899 on Build GRCm38
UniGene ID
Mm.41393
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027249.3 NP_081525.1 4 813
GenBank mRNA BC021945.1 - 4 784

View Full Product Details