Gene Transcripts

Gene Symbol
Trav5-4
Entrez Gene ID
21507
Gene Name
T cell receptor alpha variable 5-4
Gene Aliases
Tcra-V13.1
Chromosome Location
Chr. 14: 53704007 - 53704514 on Build GRCm38
UniGene ID
Mm.425911
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA M38681.1 - 208

View Full Product Details