Gene Transcripts

Gene Symbol
Adnp
Entrez Gene ID
64622
Gene Name
activity-dependent neuroprotector homeobox
Gene Aliases
Chromosome Location
Chr. 3: 164937188 - 164964819 on Build Rnor_6.0
UniGene ID
Rn.48777
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022681.2 NP_073172.2 4 1577
GenBank mRNA AF234680.1 - 1 597

View Full Product Details