Gene Transcripts

Gene Symbol
Ropn1
Entrez Gene ID
288053
Gene Name
rhophilin associated tail protein 1
Gene Aliases
Chromosome Location
Chr. 11: 69326785 - 69355788 on Build Rnor_6.0
UniGene ID
Rn.127153
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025628.1 NP_001020799.1 5 647
XM_006248431.2 XP_006248493.1 5 590
GenBank mRNA BC099132.1 - 5 647

View Full Product Details