Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_003348.3 | NP_003339.1 | 4 | 1343 | |
GenBank mRNA | AK098233.1 | - | 1 | 1778 | |
BC000396.2 | - | 4 | 1069 | 2822013 | |
BC003365.1 | - | 4 | 1004 | 2900313 | |
BC108704.1 | - | 4 | 1038 | 6503770 | |
D83004.1 | - | 4 | 1040 | ||
FJ224354.1 | - | 4 | 1040 |