Gene Transcripts

Gene Symbol
Siglec10
Entrez Gene ID
292844
Gene Name
sialic acid binding Ig-like lectin 10
Gene Aliases
Siglecg
Chromosome Location
Chr. 1: 98512663 - 98521740 on Build Rnor_6.0
UniGene ID
Rn.29166
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106250.1 NP_001099720.1 8 1798
XM_006228948.2 XP_006229010.1 11 1957
GenBank mRNA BC168854.1 - 6 1031

View Full Product Details