Gene Transcripts

Gene Symbol
XK
Entrez Gene ID
7504
Gene Name
X-linked Kx blood group
Gene Aliases
KX, MCLDS, NA, NAC, X1k, XKR1
Chromosome Location
Chr. X: 37685766 - 37732130 on Build GRCh38
UniGene ID
Hs.78919
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021083.2 NP_066569.1 3 1039
GenBank mRNA AY534238.1 - 3 1163
BC036019.1 - 3 1080 5272840
Z32684.2 - 3 1039

View Full Product Details