Gene Transcripts

Gene Symbol
Chchd5
Entrez Gene ID
296147
Gene Name
coiled-coil-helix-coiled-coil-helix domain containing 5
Gene Aliases
Chromosome Location
Chr. 3: 121659967 - 121664286 on Build Rnor_6.0
UniGene ID
Rn.8658
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106509.1 NP_001099979.1 2 242
XM_006234969.2 XP_006235031.1 2 287
GenBank mRNA BC158612.1 - 2 144

View Full Product Details